Please login to your online account to display your discounted pricing

3-Aminophenylboronic acid hemisulfate, 98%, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  104300010

 View more versions of this product

Catalog No. AC104300010

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Formula Weight 185.99
Physical Form Crystalline Powder
Quantity 1g
Color White to slightly beige
CAS Max % 100.0
Infrared Spectrum Authentic
CAS 66472-86-4
Synonym 3-Aminobenzeneboronic acid hemisulfate
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Molecular Formula C6H8BNO2·H2O4S
CAS Min % 97.5
Packaging Glass bottle
Linear Formula H2NC6H4B(OH)2·1/2H2SO4
Health Hazard 1 Warning
Chemical Name or Material 3-Aminophenylboronic acid hemisulfate
Assay Percent Range 98% (Assay acid ≥ 97.5%)
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
MDL Number MFCD00013111

Specifications

Chemical Name or Material 3-Aminophenylboronic acid hemisulfate
CAS 66472-86-4
CAS Min % 97.5
CAS Max % 100.0
Physical Form Crystalline Powder
Assay ≥97.5%
Assay Percent Range 98% (Assay acid ≥ 97.5%)
Molecular Formula C6H8BNO2·H2O4S
Linear Formula H2NC6H4B(OH)2·1/2H2SO4
Formula Weight 185.99
MDL Number MFCD00013111
Synonym 3-Aminobenzeneboronic acid hemisulfate
Accessories

Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

SDS & Safety Documents