missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Aminophenylacetylene, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC364720250

 View more versions of this product

Catalog No. AC364720250


 

Specifications Description & Specifications

Specifications

3-Aminophenylacetylene
54060-30-9
100.0
Glass bottle
1.04g/mL
Liquid
98%
C8H7N
117.15
3-Ethynylaniline
98%
97.5
Authentic
1.6140 to 1.6180 (20°C, 589nm)
Clear yellow to brown
≥97.5%
25g
HN(C6H4)CCH
MFCD00014779

Chemical Identifiers

54060-30-9
100.0
C8H7N
117.15
3-Ethynylaniline
≥97.5%
97.5
98%
HN(C6H4)CCH
MFCD00014779
3-Aminophenylacetylene, 98%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Flammable liquid and vapor.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning