Learn More
3-Aminophenol, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 146100050
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
3-Aminophenol | |
591-27-5 | |
100.0 | |
Gray or Pink to White | |
155°C | |
98.5% min. (GC) | |
C6H7NO | |
MFCD00007786 | |
13, 401 | |
m-aminophenol, m-hydroxyaniline, 3-hydroxyaniline, phenol, 3-amino, 1-amino-3-hydroxybenzene, 3-amino-1-hydroxybenzene, fouramine eg, futramine eg, fourrine eg, pelagol eg | |
CWLKGDAVCFYWJK-UHFFFAOYSA-N | |
3-aminophenol | |
11568 | |
109.13 | |
Crystalline Powder |
99% | |
98.5 | |
121.0°C to 124.0°C | |
164°C (15.0 hPa) | |
Authentic | |
Glass bottle | |
H2NC6H4OH | |
5 g | |
15, 456 | |
Solubility in water: 35g/L (20°C). Other solubilities: freely soluble in hot water, freely soluble in alcohol, ether and amyl alcohol, slightly soluble in benzene, very slightly soluble in petroleum ether | |
NC1=CC=CC(O)=C1 | |
109.13 | |
CHEBI:28924 | |
99% |
Chemical Identifiers
591-27-5 | |
109.13 | |
CWLKGDAVCFYWJK-UHFFFAOYSA-N | |
11568 | |
3-aminophenol |
C6H7NO | |
MFCD00007786 | |
m-aminophenol, m-hydroxyaniline, 3-hydroxyaniline, phenol, 3-amino, 1-amino-3-hydroxybenzene, 3-amino-1-hydroxybenzene, fouramine eg, futramine eg, fourrine eg, pelagol eg | |
CHEBI:28924 | |
NC1=CC=CC(O)=C1 |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Harmful if inhaled.
Toxic to aquatic life with long lasting effects.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
GHS Signal Word: Warning
EINECSNumber : 209-711-2
RTECSNumber : SJ4900000
TSCA : TSCA
RUO – Research Use Only