missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Aminophenol, 98+%

Manufacturer:  Alfa Aesar™ A119430B

 View more versions of this product

Catalog No. AAA119430B



Description

Description

Applications
A chemical used in the preparation of inhibitors of mammalian carbonic anhydrase isoforms. It is also used in the preparation of quinoline tethered fluorescent carbon dots for regulated anticancer discovery. It is employed in the production of TB drugs, salicylic acid, amino stabilizer and developers. 3-Aminophenol has been used as a stabilizer of chlorine-containing thermoplastics, although its principal use is as an intermediate in the production of 4-amino-2-hydroxybenzoic acid this isomer is also employed as a hair colorant and as a coupler molecule in hair dyes.

Solubility
Soluble in water (35 mg/ml at 20°C), ether, alcohol, amyl alcohol, and benzene (slightly).

Notes
Store away from oxidizing agents. Keep the container tightly closed. Store in cool, dry conditions in well ventilated place.
Specifications

Specifications

3-Aminophenol
Characteristic
122°C to 126°C
6.8
C6H7NO
UN2512
14,460
CWLKGDAVCFYWJK-UHFFFAOYSA-N
3-aminophenol
11568
109.13
178°C (352°F)
164°C (11mmHg)
1000g
591-27-5
MFCD00007786
636059
m-aminophenol, m-hydroxyaniline, 3-hydroxyaniline, phenol, 3-amino, 1-amino-3-hydroxybenzene, 3-amino-1-hydroxybenzene, fouramine eg, futramine eg, fourrine eg, pelagol eg
C1=CC(=CC(=C1)O)N
109.128
CHEBI:28924
≥98%

Chemical Identifiers

591-27-5
109.128
CWLKGDAVCFYWJK-UHFFFAOYSA-N
11568
3-aminophenol
C6H7NO
MFCD00007786
m-aminophenol, m-hydroxyaniline, 3-hydroxyaniline, phenol, 3-amino, 1-amino-3-hydroxybenzene, 3-amino-1-hydroxybenzene, fouramine eg, futramine eg, fourrine eg, pelagol eg
CHEBI:28924
C1=CC(=CC(=C1)O)N
Safety and Handling

Safety and Handling

GHS H Statement
H302-H332
Harmful if swallowed.
Harmful if inhaled.

GHS P Statement
P273-P302+P352
Avoid release to the environment.
IF ON SKIN: Wash with plenty of soap and water.

Warning

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: AMINOPHENOLS (O-;M-;P-)

RTECSNumber : SJ4900000

TSCA : TSCA

SDS Documents