Learn More
3-Aminopentane, 98+%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 183700050
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
3-Aminopentane | |
98.0 | |
0.7400g/mL | |
2°C | |
98% min. (GC) | |
C5H13N | |
(C2H5)2CHNH2 | |
5g | |
0.74 | |
Solubility in water: soluble. | |
CCC(N)CC | |
87.17 | |
CHEBI:84248 | |
98+% |
616-24-0 | |
100.0 | |
89.0°C to 91.0°C | |
Authentic | |
Glass bottle | |
1.4055 | |
MFCD00008096 | |
04, 179 | |
3-aminopentane, 3-pentanamine, 1-ethylpropylamine, 3-pentylamine, propylamine, 1-ethyl, 3-amylamine, unii-3n2it605hv, 1-ethyl-propylamine, pent-3-ylamine, 3-amino pentane | |
PQPFFKCJENSZKL-UHFFFAOYSA-N | |
pentan-3-amine | |
12019 | |
87.16 |
Chemical Identifiers
616-24-0 | |
87.17 | |
PQPFFKCJENSZKL-UHFFFAOYSA-N | |
12019 | |
pentan-3-amine |
C5H13N | |
MFCD00008096 | |
3-aminopentane, 3-pentanamine, 1-ethylpropylamine, 3-pentylamine, propylamine, 1-ethyl, 3-amylamine, unii-3n2it605hv, 1-ethyl-propylamine, pent-3-ylamine, 3-amino pentane | |
CHEBI:84248 | |
CCC(N)CC |
Safety and Handling
GHS H Statement
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
GHS Signal Word: Danger
EINECSNumber : 210-471-6
RUO â Research Use Only