Learn More
3-Aminooxetane, 95%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 451215000
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
3-Aminooxetane | |
99°C | |
94% min. (GC) | |
500mg | |
OJEOJUQOECNDND-UHFFFAOYSA-N | |
oxetan-3-amine | |
9833923 | |
95% |
21635-88-1 | |
24°C | |
C3H7NO | |
3-oxetanamine, 3-aminooxetane, oxetan-3-ylamine, 3-oxetanamin, 3-amino-oxetane, oxetane-3-amine, oxetan-3-yl amine, pubchem20414, acmc-1cgam | |
C1C(CO1)N | |
73.09 | |
73.09 |
Chemical Identifiers
21635-88-1 | |
73.09 | |
3-oxetanamine, 3-aminooxetane, oxetan-3-ylamine, 3-oxetanamin, 3-amino-oxetane, oxetane-3-amine, oxetan-3-yl amine, pubchem20414, acmc-1cgam | |
oxetan-3-amine |
C3H7NO | |
OJEOJUQOECNDND-UHFFFAOYSA-N | |
9833923 | |
C1C(CO1)N |
Safety and Handling
GHS H Statement
Flammable liquid and vapour.
Toxic if swallowed.
Causes serious eye damage.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water f
GHS Signal Word: Danger
RUO â Research Use Only