Learn More
3-Aminooxetane, 95%, Thermo Scientific Chemicals
$284.68 - $472.18
Chemical Identifiers
CAS | 21635-88-1 |
---|---|
Molecular Formula | C3H7NO |
Molecular Weight (g/mol) | 73.095 |
MDL Number | MFCD08544363 |
InChI Key | OJEOJUQOECNDND-UHFFFAOYSA-N |
Synonym | 3-oxetanamine, 3-aminooxetane, oxetan-3-ylamine, 3-oxetanamin, 3-amino-oxetane, oxetane-3-amine, oxetan-3-yl amine, pubchem20414, acmc-1cgam |
PubChem CID | 9833923 |
IUPAC Name | oxetan-3-amine |
SMILES | C1C(CO1)N |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH57248MD
|
Thermo Scientific Chemicals
H57248MD |
250 mg |
Each for $284.68
|
|
|||||
AAH57248ME
|
Thermo Scientific Chemicals
H57248ME |
500 mg |
Each for $472.18
|
|
|||||
Description
It is used in medicine.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is used in medicine.
Solubility
Slightly soluble in water
Notes
Air sensitive. Store in a cool ventilated place. Avoid strong oxidizing materials and heat.
Chemical Identifiers
21635-88-1 | |
73.095 | |
OJEOJUQOECNDND-UHFFFAOYSA-N | |
9833923 | |
C1C(CO1)N |
C3H7NO | |
MFCD08544363 | |
3-oxetanamine, 3-aminooxetane, oxetan-3-ylamine, 3-oxetanamin, 3-amino-oxetane, oxetane-3-amine, oxetan-3-yl amine, pubchem20414, acmc-1cgam | |
oxetan-3-amine |
Specifications
21635-88-1 | |
1.05 g/mL | |
24°C (75°F) | |
MFCD08544363 | |
UN1993 | |
3-oxetanamine, 3-aminooxetane, oxetan-3-ylamine, 3-oxetanamin, 3-amino-oxetane, oxetane-3-amine, oxetan-3-yl amine, pubchem20414, acmc-1cgam | |
OJEOJUQOECNDND-UHFFFAOYSA-N | |
oxetan-3-amine | |
9833923 | |
95% | |
3-Aminooxetane |
Colorless to Yellow | |
99°C | |
C3H7NO | |
250 mg | |
Air sensitive | |
Slightly soluble in water | |
C1C(CO1)N | |
73.095 | |
73.09 | |
Liquid |
Safety and Handling
GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P233-P240-P241-P242-P243-P264b-P270-P280-P301+P310-P303+P361+P353-P305+P351+P338-P310-P330-P370+P378q-P501c
H226-H301-H318
DOTInformation : Transport Hazard Class: 3; Packaging Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
TSCA : No
storageNote1 : -30° to -10°C
Recommended Storage : Store at -20°C; Store under Argon
RUO – Research Use Only