missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3-Aminooxetane-3-carboxylic acid, 95%, Thermo Scientific Chemicals
$494.29 - $901.74
Chemical Identifiers
CAS | 138650-24-5 |
---|---|
Molecular Formula | C4H7NO3 |
Molecular Weight (g/mol) | 117.104 |
MDL Number | MFCD11035888 |
InChI Key | LXRNYRAHXDUNKR-UHFFFAOYSA-N |
Synonym | 3-oxetanecarboxylic acid, 3-amino, 3-amino-oxetane-3-carboxylic acid, acmc-20a7wi, 3-amino-3-oxetanecarboxylic acid, 3-aminooxetane-3-carboxylic acid |
PubChem CID | 15024256 |
IUPAC Name | 3-aminooxetane-3-carboxylic acid |
SMILES | C1C(CO1)(C(=O)O)N |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH57291MD
|
Thermo Scientific Chemicals
H57291MD |
250 mg |
Each for $494.29
|
|
|||||
AAH57291ME
|
Thermo Scientific Chemicals
H57291ME |
500 mg |
Each for $901.74
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
138650-24-5 | |
117.104 | |
LXRNYRAHXDUNKR-UHFFFAOYSA-N | |
15024256 | |
C1C(CO1)(C(=O)O)N |
C4H7NO3 | |
MFCD11035888 | |
3-oxetanecarboxylic acid, 3-amino, 3-amino-oxetane-3-carboxylic acid, acmc-20a7wi, 3-amino-3-oxetanecarboxylic acid, 3-aminooxetane-3-carboxylic acid | |
3-aminooxetane-3-carboxylic acid |
Specifications
138650-24-5 | |
White | |
MFCD11035888 | |
3-oxetanecarboxylic acid, 3-amino, 3-amino-oxetane-3-carboxylic acid, acmc-20a7wi, 3-amino-3-oxetanecarboxylic acid, 3-aminooxetane-3-carboxylic acid | |
C1C(CO1)(C(=O)O)N | |
117.104 | |
117.1 | |
Powder |
205°C to 210°C | |
C4H7NO3 | |
250 mg | |
LXRNYRAHXDUNKR-UHFFFAOYSA-N | |
3-aminooxetane-3-carboxylic acid | |
15024256 | |
95% | |
3-Aminooxetane-3-carboxylic acid |
Safety and Handling
GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P264b-P270-P280i-P301+P312-P305+P351+P338-P330-P501c
H302-H319
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only