Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Aminonoradamantane hydrochloride, 95%

Chemical Identifiers

Chemical Name or Material 3-Aminonoradamantane hydrochloride
CAS 86128-83-8
Formula Weight 173.68
MDL Number MFCD00213499
Synonym 3-Noradamantanamine hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6612003

 
alfa aesar™
H6612003
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH66120MD

 
alfa aesar™
H66120MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 173.68
Chemical Name or Material 3-Aminonoradamantane hydrochloride
CAS 86128-83-8
Assay 95%
Synonym 3-Noradamantanamine hydrochloride
Physical Form Powder
Color White to off-white
Melting Point 350°C
Quantity 1g
MDL Number MFCD00213499