missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Aminomethyltetrahydrofuran, 97%, ACROS Organics™

Manufacturer:  Acros Organics AC433000050

 View more versions of this product

Catalog No. AC433000050



Specifications

Specifications

3-Aminomethyltetrahydrofuran
Glass bottle
Colorless
165253-31-6
100.0
C5H11NO
tetrahydrofuran-3-yl methanamine, tetrahydro-3-furanmethanamine, tetrahydrofuran-3-ylmethylamine, 3-furanmethanamine, tetrahydro, 1-tetrahydrofuran-3-ylmethanamine, c-tetrahydro-furan-3-yl-methylamine, 3-aminomethyl oxolane, 3-aminomethyl tetrahydrofuran, tetrahydrofur-3-yl methylamine, c-tetrahydrofuran-3-yl methylamine
C1COCC1CN
101.149
101.15
97%
Authentic
65.0°C (15.0 Torr)
5g
96.0
97%
MFCD08234925
CINJIXGRSTYIHP-UHFFFAOYSA-N
oxolan-3-ylmethanamine
10898660
Solution

Chemical Identifiers

165253-31-6
101.149
CINJIXGRSTYIHP-UHFFFAOYSA-N
10898660
C1COCC1CN
C5H11NO
MFCD08234925
tetrahydrofuran-3-yl methanamine, tetrahydro-3-furanmethanamine, tetrahydrofuran-3-ylmethylamine, 3-furanmethanamine, tetrahydro, 1-tetrahydrofuran-3-ylmethanamine, c-tetrahydro-furan-3-yl-methylamine, 3-aminomethyl oxolane, 3-aminomethyl tetrahydrofuran, tetrahydrofur-3-yl methylamine, c-tetrahydrofuran-3-yl methylamine
oxolan-3-ylmethanamine
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Flammable liquid and vapour.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

SDS Documents