Please login to your online account to display your discounted pricing

3-(Aminomethyl)pyridine, 99+%, ACROS Organics™


Quantity 1L
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC104080010

 View more versions of this product

Catalog No. AC104080010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Liquid
Assay ≥99.0%
Formula Weight 108.14
Quantity 1L
Color Clear colorless to slightly yellow
TSCA TSCA
EINECS Number 223-091-0
Density 1.0600g/mL
CAS Min % 99.0
CAS Max % 100.0
Health Hazard 1 GHS Signal Word: Danger
CAS 3731-52-0
Health Hazard 2 GHS H Statement:
Causes serious eye damage.
Harmful in contact with skin.
Causes severe skin burns and eye damage.
Harmful if swallowed.
Chemical Name or Material 3-(Aminomethyl)pyridine
Name Note 99+%
Molecular Formula C6H8N2
MDL Number MFCD00006412
Refractive Index 1.5500 to 1.5520 (20°C, 589nm)
Assay Percent Range 99%
Packaging Glass bottle
Infrared Spectrum Authentic
Synonym 3-Picolylamine
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material 3-(Aminomethyl)pyridine
Name Note 99+%
CAS 3731-52-0
CAS Min % 99.0
CAS Max % 100.0
Assay ≥99.0%
Assay Percent Range 99%
Molecular Formula C6H8N2
Formula Weight 108.14
MDL Number MFCD00006412
Synonym 3-Picolylamine
Accessories

GHS Signal Word: Danger

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye damage.
Harmful in contact with skin.
Causes severe skin burns and eye damage.
Harmful if swallowed.

SDS & Safety Documents