Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-(Aminomethyl)oxetane, 96%

Chemical Identifiers

Chemical Name or Material 3-(Aminomethyl)oxetane
CAS 6246-05-5
Formula Weight 87.12
MDL Number MFCD12547197
Synonym 3-Oxetanemethylamine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH66510MD

  SDS
alfa aesar™
H66510MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 87.12
CAS 6246-05-5
Color Colorless to pale yellow
Physical Form Liquid
Assay 96%
Quantity 250mg
MDL Number MFCD12547197
Synonym 3-Oxetanemethylamine
Chemical Name or Material 3-(Aminomethyl)oxetane
Sensitivity Air Sensitive