missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-(Aminomethyl)benzeneboronic acid hydrochloride, 96%

Manufacturer:  Alfa Aesar™ H5328403

 View more versions of this product

Catalog No. AAH5328403



Description

Description

Applications
3-(Aminomethyl)benzeneboronic acid hydrochloride is used as intermediates. Hydroxybenzene boronic acids are involved in suzuki miyaura reactions. It is a arylboronic acid. 3-Fluoro-4-hydroxybenzeneboronic acid, can be an effective catalyst for amidation and esterification of carboxylic acids.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents. Refrigerate.
Specifications

Specifications

3-(Aminomethyl)benzeneboronic acid hydrochloride
1g
C7H11BClNO2
3-aminomethyl phenyl boronic acid hydrochloride, 3-aminomethyl phenylboronic acid hydrochloride, 3-aminomethylphenylboronic acid hydrochloride, 3-aminomethylphenyl boronic acid hydrochloride, 3-aminomethylphenylboronic acid hcl, 3-aminomethylphenyl boronic acid hcl, 3-aminomethyl phenyl boronic acid hydrochloride, boronic acid, 3-aminomethyl phenyl-, hydrochloride, 3-aminomethylphenyl boronic acid hcl, 3-aminomethylphenylboronic acid, hcl
B(C1=CC(=CC=C1)CN)(O)O.Cl
187.43
187.43
248°C to 250°C
352525-94-1
MFCD02093050
NPTBTFRGCBFYPZ-UHFFFAOYSA-N
[3-(aminomethyl)phenyl]boronic acid;hydrochloride
16427085
96%

Chemical Identifiers

352525-94-1
187.43
NPTBTFRGCBFYPZ-UHFFFAOYSA-N
16427085
B(C1=CC(=CC=C1)CN)(O)O.Cl
C7H11BClNO2
MFCD02093050
3-aminomethyl phenyl boronic acid hydrochloride, 3-aminomethyl phenylboronic acid hydrochloride, 3-aminomethylphenylboronic acid hydrochloride, 3-aminomethylphenyl boronic acid hydrochloride, 3-aminomethylphenylboronic acid hcl, 3-aminomethylphenyl boronic acid hcl, 3-aminomethyl phenyl boronic acid hydrochloride, boronic acid, 3-aminomethyl phenyl-, hydrochloride, 3-aminomethylphenyl boronic acid hcl, 3-aminomethylphenylboronic acid, hcl
[3-(aminomethyl)phenyl]boronic acid;hydrochloride
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

SDS Documents