missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Aminomethyl-2-bromothiophene, 95%

$43.81 - $43.81

Chemical Identifiers

CAS 157664-47-6
Molecular Formula C5H6BrNS
Molecular Weight (g/mol) 192.074
MDL Number MFCD02180914
InChI Key SLKZUJBSIKGHFU-UHFFFAOYSA-N
Synonym 2-bromo-3-thienyl methylamine, 2-bromothiophen-3-yl methanamine, 2-bromo-3-thiophenemethylamine, 3-thiophenemethanamine,2-bromo, 1-2-bromothiophen-3-yl methanamine, 2-bromothien-3-yl methylamine hydrochloride, pubchem5276, h-val-ssna, bestipharma 583-851, 3-aminomethyl-2-bromothiophene
PubChem CID 2735586
IUPAC Name (2-bromothiophen-3-yl)methanamine
SMILES C1=CSC(=C1CN)Br
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH66245MD View Documents Alfa Aesar™
H66245MD
250mg Each for $43.81
 
Specifications

Chemical Identifiers

157664-47-6
192.074
SLKZUJBSIKGHFU-UHFFFAOYSA-N
2735586
C1=CSC(=C1CN)Br
C5H6BrNS
MFCD02180914
2-bromo-3-thienyl methylamine, 2-bromothiophen-3-yl methanamine, 2-bromo-3-thiophenemethylamine, 3-thiophenemethanamine,2-bromo, 1-2-bromothiophen-3-yl methanamine, 2-bromothien-3-yl methylamine hydrochloride, pubchem5276, h-val-ssna, bestipharma 583-851, 3-aminomethyl-2-bromothiophene
(2-bromothiophen-3-yl)methanamine

Specifications

1.657g/mL
Air Sensitive
250mg
C5H6BrNS
UN2810
SLKZUJBSIKGHFU-UHFFFAOYSA-N
(2-bromothiophen-3-yl)methanamine
2735586
Liquid
3-Aminomethyl-2-bromothiophene
1.6100
Colorless to Yellow
157664-47-6
MFCD02180914
2-bromo-3-thienyl methylamine, 2-bromothiophen-3-yl methanamine, 2-bromo-3-thiophenemethylamine, 3-thiophenemethanamine,2-bromo, 1-2-bromothiophen-3-yl methanamine, 2-bromothien-3-yl methylamine hydrochloride, pubchem5276, h-val-ssna, bestipharma 583-851, 3-aminomethyl-2-bromothiophene
C1=CSC(=C1CN)Br
192.074
192.08
95%
Safety and Handling

Safety and Handling

GHS H Statement
H301-H318-H315-H335
Toxic if swallowed.
Causes serious eye damage.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.