missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Aminocarbonylphenylboronic acid, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC444890050

 View more versions of this product

Catalog No. AC444890050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

3-Aminocarbonylphenylboronic acid
Glass bottle
232.0°C to 238.0°C
351422-73-6
C7H8BNO3
WDGWHKRJEBENCE-UHFFFAOYSA-N
(3-carbamoylphenyl)boronic acid
2737814
Solid
96% min. consistent with structure (1H-NMR)
White
5g
96% min. (HPLC)
3-aminocarbonyl benzeneboronic acid, 3-aminocarbonyl phenyl boronic acid, 3-aminocarbonyl phenylboronic acid, 3-aminocarbonylphenyl boronic acid, 3-aminocarbonylphenylboronic acid, 3-aminocarbonyphenylboronic acid, 3-carbamoylbenzeneboronic acid, 3-carbamoylphenyl boronic acid, 3-carboxamidephenylboronic acid, benzamide-3-boronic acid
B(C1=CC(=CC=C1)C(=O)N)(O)O
164.96
164.96
97%

Chemical Identifiers

351422-73-6
164.96
3-aminocarbonyl benzeneboronic acid, 3-aminocarbonyl phenyl boronic acid, 3-aminocarbonyl phenylboronic acid, 3-aminocarbonylphenyl boronic acid, 3-aminocarbonylphenylboronic acid, 3-aminocarbonyphenylboronic acid, 3-carbamoylbenzeneboronic acid, 3-carbamoylphenyl boronic acid, 3-carboxamidephenylboronic acid, benzamide-3-boronic acid
(3-carbamoylphenyl)boronic acid
C7H8BNO3
WDGWHKRJEBENCE-UHFFFAOYSA-N
2737814
B(C1=CC(=CC=C1)C(=O)N)(O)O
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present

GHS Signal Word: Warning

SDS Documents