missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3-Aminocarbonylphenylboronic acid, 97%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC444890010
Specifications
3-Aminocarbonylphenylboronic acid | |
Glass bottle | |
232.0°C to 238.0°C | |
351422-73-6 | |
C7H8BNO3 | |
WDGWHKRJEBENCE-UHFFFAOYSA-N | |
(3-carbamoylphenyl)boronic acid | |
2737814 | |
Solid |
96% min. consistent with structure (1H-NMR) | |
White | |
1g | |
96% min. (HPLC) | |
3-aminocarbonyl benzeneboronic acid, 3-aminocarbonyl phenyl boronic acid, 3-aminocarbonyl phenylboronic acid, 3-aminocarbonylphenyl boronic acid, 3-aminocarbonylphenylboronic acid, 3-aminocarbonyphenylboronic acid, 3-carbamoylbenzeneboronic acid, 3-carbamoylphenyl boronic acid, 3-carboxamidephenylboronic acid, benzamide-3-boronic acid | |
B(C1=CC(=CC=C1)C(=O)N)(O)O | |
164.96 | |
164.96 | |
97% |
Chemical Identifiers
351422-73-6 | |
164.96 | |
3-aminocarbonyl benzeneboronic acid, 3-aminocarbonyl phenyl boronic acid, 3-aminocarbonyl phenylboronic acid, 3-aminocarbonylphenyl boronic acid, 3-aminocarbonylphenylboronic acid, 3-aminocarbonyphenylboronic acid, 3-carbamoylbenzeneboronic acid, 3-carbamoylphenyl boronic acid, 3-carboxamidephenylboronic acid, benzamide-3-boronic acid | |
(3-carbamoylphenyl)boronic acid |
C7H8BNO3 | |
WDGWHKRJEBENCE-UHFFFAOYSA-N | |
2737814 | |
B(C1=CC(=CC=C1)C(=O)N)(O)O |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present
GHS Signal Word: Warning