Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Aminocarbonyl-5-nitrobenzoic acid, 97%

$129.73 - $387.24

Chemical Identifiers

Chemical Name or Material 3-Aminocarbonyl-5-nitrobenzoic acid
CAS 90196-48-8
Molecular Formula C8H6N2O5
Formula Weight 210.15
MDL Number MFCD00127687
Synonym 3-Carboxy-5-nitrobenzamide; 5-Nitroisophthalamic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2622706

  SDS
alfa aesar™
H2622706
5g Each for $129.73

AAH2622714

  SDS
alfa aesar™
H2622714
25g Each for $387.24
Description & Specifications

Specifications

Formula Weight 210.15
Chemical Name or Material 3-Aminocarbonyl-5-nitrobenzoic acid
Melting Point 239° to 242°C
Quantity 5g
Assay 97%
Molecular Formula C8H6N2O5
MDL Number MFCD00127687
CAS 90196-48-8
Synonym 3-Carboxy-5-nitrobenzamide; 5-Nitroisophthalamic acid
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.