missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Aminobiphenyl hydrochloride

$158.57 - $707.57

Chemical Identifiers

CAS 2113-55-5
Molecular Formula C12H11N·HCl
Formula Weight 205.69
MDL Number MFCD00068358
Synonym 3-Biphenylamine hydrochloride; 3-Phenylaniline hydrochloride
Chemical Name or Material 3-Aminobiphenyl hydrochloride
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH5102706 SDS Alfa Aesar™
H5102706
5g Each for $707.57
AAH5102703 SDS Alfa Aesar™
H5102703
1g Each for $158.57
Specifications Description & Specifications

Chemical Identifiers

2113-55-5
205.69
3-Biphenylamine hydrochloride; 3-Phenylaniline hydrochloride
C12H11N·HCl
MFCD00068358
3-Aminobiphenyl hydrochloride

Specifications

2113-55-5
205.69
MFCD00068358
3-Biphenylamine hydrochloride; 3-Phenylaniline hydrochloride
C12H11N·HCl
5g
Odorless
3-Aminobiphenyl hydrochloride

Safety and Handling

GHS H Statement
H351-H302
Suspected of causing cancer.
Harmful if swallowed.

GHS P Statement
P280h
Wear protective gloves/clothing.

Warning