Learn More
3-Aminobenzyl alcohol, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 184570050
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
3-Aminobenzyl alcohol | |
96.0 | |
91.0°C to 95.0°C | |
Authentic | |
Glass bottle | |
H2NC6H4CH2OH | |
5 g | |
3-aminobenzyl alcohol, 3-aminophenyl methanol, m-aminobenzyl alcohol, 3-aminobenzylalcohol, 3-hydroxymethyl aniline, benzenemethanol, 3-amino, benzyl alcohol, m-amino, 3-aminobenzenemethanol, unii-kn96abq0sp, m-aminophenylcarbinol | |
NC1=CC=CC(CO)=C1 | |
123.16 | |
123.15 | |
Fine Crystalline Powder |
1877-77-6 | |
100.0 | |
Beige or Beige-Gray to Brown | |
96% min. (GC) | |
C7H9NO | |
MFCD00007817 | |
13, 619 | |
OJZQOQNSUZLSMV-UHFFFAOYSA-N | |
(3-aminophenyl)methanol | |
80293 | |
97% |
Chemical Identifiers
1877-77-6 | |
123.16 | |
OJZQOQNSUZLSMV-UHFFFAOYSA-N | |
80293 | |
NC1=CC=CC(CO)=C1 |
C7H9NO | |
MFCD00007817 | |
3-aminobenzyl alcohol, 3-aminophenyl methanol, m-aminobenzyl alcohol, 3-aminobenzylalcohol, 3-hydroxymethyl aniline, benzenemethanol, 3-amino, benzyl alcohol, m-amino, 3-aminobenzenemethanol, unii-kn96abq0sp, m-aminophenylcarbinol | |
(3-aminophenyl)methanol |
Safety and Handling
GHS H Statement
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and kee
GHS Signal Word: Warning
EINECSNumber : 217-514-8
RUO – Research Use Only