Learn More
3-Aminobenzotrifluoride, 98%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC103190025
Specifications
3-Aminobenzotrifluoride | |
85°C | |
Glass bottle | |
1.303 | |
Yellow | |
2.5L | |
98-16-8 | |
97.5% min. (GC) | |
H2NC6H4CF3 | |
12, 870 | |
Solubility in water: 5g/L in water (20°C). Other solubilities: soluble in ethanol,ether and diethyl ether | |
C1=CC(=CC(=C1)N)C(F)(F)F | |
161.13 | |
161.13 | |
98% |
1.3030g/mL | |
Authentic | |
1.4790 to 1.4810 | |
187.0°C to 193.0°C | |
5.0°C to 6.0°C | |
98% | |
98% | |
C7H6F3N | |
MFCD00007797 | |
3-aminobenzotrifluoride, 3-aminotrifluorotoluene, 3-trifluoromethyl aniline, 3-trifluoromethyl benzenamine, benzenamine, 3-trifluoromethyl, m-abtf, m-amino trifluorotoluene, m-aminobenzotrifluoride, m-trifluoromethyl aniline, m-trifluoromethylaniline | |
VIUDTWATMPPKEL-UHFFFAOYSA-N | |
3-(trifluoromethyl)aniline | |
7375 | |
Liquid |
Chemical Identifiers
98-16-8 | |
161.13 | |
VIUDTWATMPPKEL-UHFFFAOYSA-N | |
7375 | |
C1=CC(=CC(=C1)N)C(F)(F)F |
C7H6F3N | |
MFCD00007797 | |
3-aminobenzotrifluoride, 3-aminotrifluorotoluene, 3-trifluoromethyl aniline, 3-trifluoromethyl benzenamine, benzenamine, 3-trifluoromethyl, m-abtf, m-amino trifluorotoluene, m-aminobenzotrifluoride, m-trifluoromethyl aniline, m-trifluoromethylaniline | |
3-(trifluoromethyl)aniline |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Fatal if inhaled.
Causes skin irritation.
Causes serious eye damage.
May cause respiratory irritation.
May cause damage to organs through prolo
GHS P Statement
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN: Wash w
GHS Signal Word: Danger
EINECSNumber : 202-643-4
RTECSNumber : XU9180000
TSCA : TSCA
Recommended Storage : This product may darken on storage