Please login to your online account to display your discounted pricing

3-Aminobenzotrifluoride 98%, ACROS Organics™


Quantity 5mL
Packaging Glass bottle

Manufacturer: thermo fisher scientific  103190050

 View more versions of this product

Catalog No. AC103190050

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥98%
Physical Form Liquid
Formula Weight 161.13
Quantity 5mL
Color Clear light yellow
Beilstein 12, 870
EINECS Number 202-643-4
TSCA TSCA
RTECS Number XU9180000
CAS Min % 98
Packaging Glass bottle
Name Note 98%
Linear Formula H2NC6H4CF3
Chemical Name or Material 3-Aminobenzotrifluoride
Refractive Index 1.4790 to 1.4810 (20°C, 589nm)
Assay Percent Range 98%
Molecular Formula C7H6F3N
MDL Number MFCD00007797
CAS 98-16-8
Synonym 3-(Trifluoromethyl)aniline
Health Hazard 1 GHS Signal Word: Warning
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
May cause damage to organs through prolonged or repeated exposure.
Fatal if inhaled.
Harmful in contact with skin.
Causes skin irritation.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic
Storage Information This product may darken on storage

Specifications

Chemical Name or Material 3-Aminobenzotrifluoride
Name Note 98%
CAS 98-16-8
CAS Min % 98
Assay ≥98%
Assay Percent Range 98%
Molecular Formula C7H6F3N
Linear Formula H2NC6H4CF3
Formula Weight 161.13
MDL Number MFCD00007797
Synonym 3-(Trifluoromethyl)aniline
Accessories

GHS Signal Word: Warning

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
May cause damage to organs through prolonged or repeated exposure.
Fatal if inhaled.
Harmful in contact with skin.
Causes skin irritation.

SDS & Safety Documents