Please login to your online account to display your discounted pricing

3-Aminobenzophenone 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 3-Aminobenzophenone
CAS 2835-78-1
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C13H11NO
Linear Formula C6H5C(O)C6H4NH2
Formula Weight 197.24
MDL Number MFCD00017103
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC392860050

  SDS
thermo fisher scientific
AC392860050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC392860010

  SDS
thermo fisher scientific
AC392860010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC392860250

  SDS
thermo fisher scientific
AC392860250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥97.5%
Physical Form Powder
Color Yellow to green
Formula Weight 197.24
Quantity 5g
Melting Point 84° to 87°C
CAS Max % 100.0
Chemical Name or Material 3-Aminobenzophenone
MDL Number MFCD00017103
Synonym (3-Aminophenyl)(phenyl)methanone
Infrared Spectrum Authentic
Packaging Glass bottle
Assay Percent Range 98%
Molecular Formula C13H11NO
Linear Formula C6H5C(O)C6H4NH2
CAS 2835-78-1
CAS Min % 97.5
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.