Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Aminobenzophenone, 97+%

$67.66 - $248.62

Chemical Identifiers

Chemical Name or Material 3-Aminobenzophenone
CAS 2835-78-1
Molecular Formula C13H11NO
Formula Weight 197.24
MDL Number MFCD00017103
Beilstein 2209413
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0896006

  SDS
alfa aesar™
L0896006
5g Each for $67.66

AAL0896014

  SDS
alfa aesar™
L0896014
25g Each for $248.62
Description & Specifications

Specifications

Formula Weight 197.24
MDL Number MFCD00017103
Melting Point 84° to 88°C
Chemical Name or Material 3-Aminobenzophenone
Molecular Formula C13H11NO
CAS 2835-78-1
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Assay ≥97%
Quantity 5g
Beilstein 2209413
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
RTECS Number DJ0170000
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.