missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Aminobenzonitrile, 99%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC173041000

 View more versions of this product

Catalog No. AC173041000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

3-Aminobenzonitrile
Authentic
288.0°C to 290.0°C
50.0°C to 54.0°C
99%
98.5
99%
H2NC6H4CN
14, 391
Solubility in water: insoluble. Other solubilities: soluble in ehtanol,ethyl ether,acetone
C1=CC(=CC(=C1)N)C#N
118.14
118.14
99%
112°C
Glass bottle
Brown
100g
2237-30-1
100.0
C7H6N2
MFCD00007756
3-amino benzonitrile, 3-amino-benzonitrile, 3-aminobenzonitril, 3-cyanoaniline, benzonitrile, 3-amino, benzonitrile, m-amino, m-aminobenzonitrile, m-anthranilonitrile, m-cyanoaniline, unii-15v3cw939x
NJXPYZHXZZCTNI-UHFFFAOYSA-N
3-aminobenzonitrile
16702
Crystalline Solid

Chemical Identifiers

2237-30-1
118.14
NJXPYZHXZZCTNI-UHFFFAOYSA-N
16702
C1=CC(=CC(=C1)N)C#N
C7H6N2
MFCD00007756
3-amino benzonitrile, 3-amino-benzonitrile, 3-aminobenzonitril, 3-cyanoaniline, benzonitrile, 3-amino, benzonitrile, m-amino, m-aminobenzonitrile, m-anthranilonitrile, m-cyanoaniline, unii-15v3cw939x
3-aminobenzonitrile
Safety and Handling

Safety and Handling

GHS H Statement
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.

GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POI

GHS Signal Word: Danger

EINECSNumber : 218-800-5

RTECSNumber : DI2454000

TSCA : TSCA

SDS Documents