Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Aminobenzoic acid hydrochloride, 98%

$42.95 - $131.09

Chemical Identifiers

Chemical Name or Material 3-Aminobenzoic acid hydrochloride
CAS 15151-51-6
Molecular Formula C7H7NO2·HCl
Formula Weight 173.6
MDL Number MFCD00012973
Beilstein 3695285
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0840714

  SDS
alfa aesar™
L0840714
25g Each for $42.95

AAL0840722

  SDS
alfa aesar™
L0840722
100g Each for $131.09
Description & Specifications

Specifications

Formula Weight 173.6
Chemical Name or Material 3-Aminobenzoic acid hydrochloride
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Assay 98%
Molecular Formula C7H7NO2·HCl
MDL Number MFCD00012973
CAS 15151-51-6
Beilstein 3695285
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECS Number DG1225000
Melting Point ≈286°C
Quantity 25g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.