Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Aminobenzhydrazide, 97%

$1,250.61 - $1,250.61

Chemical Identifiers

Chemical Name or Material 3-Aminobenzhydrazide
CAS 14062-34-1
Molecular Formula C7H9N3O
Formula Weight 151.17
MDL Number MFCD00017067
Beilstein 2090097
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1149130

  SDS
alfa aesar™
A1149130
250g Each for $1,250.61
Description & Specifications

Specifications

Formula Weight 151.17
Chemical Name or Material 3-Aminobenzhydrazide
Quantity 250g
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Assay 97%
Melting Point 90° to 94°C
Molecular Formula C7H9N3O
CAS 14062-34-1
Beilstein 2090097
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 1 Warning
MDL Number MFCD00017067
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.