Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Aminobenzenesulfonamide, tech. 80%

$58.81 - $210.88

Chemical Identifiers

Chemical Name or Material 3-Aminobenzenesulfonamide
CAS 98-18-0
Purity Grade Technical Grade
Molecular Formula C6H8N2O2S
Formula Weight 172.21
MDL Number MFCD00035781
Synonym Metanilamide; Metanilamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2543922

 
alfa aesar™
B2543922
100g Each for $58.81

AAB2543936

 
alfa aesar™
B2543936
500g Each for $210.88
Description & Specifications

Specifications

Formula Weight 172.21
Chemical Name or Material 3-Aminobenzenesulfonamide
TSCA TSCA
Purity Grade Technical Grade
Assay 80%
Melting Point 139° to 142°C
Molecular Formula C6H8N2O2S
Quantity 100g
MDL Number MFCD00035781
CAS 98-18-0
Synonym Metanilamide; Metanilamide
RTECS Number OY2200000