missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Aminobenzamidine dihydrochloride hydrate, 98%, ACROS Organics™

Chemical Identifiers

CAS 37132-68-6
Molecular Formula C7H11Cl2N3
Molecular Weight (g/mol) 208.086
MDL Number MFCD03790003
InChI Key FKFMMMVFFYPIEE-UHFFFAOYSA-N
Synonym dihydrogen 3-aminobenzenecarboximidamide dichloride
PubChem CID 74764793
IUPAC Name 3-aminobenzenecarboximidamide;hydron;dichloride
SMILES [H+].[H+].C1=CC(=CC(=C1)N)C(=N)N.[Cl-].[Cl-]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC228075000
SDS View Documents
Thermo Fisher Scientific
228075000
500mg Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC228070250
SDS View Documents
Thermo Fisher Scientific
AC228070250
25g Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

37132-68-6
208.086
FKFMMMVFFYPIEE-UHFFFAOYSA-N
74764793
[H+].[H+].C1=CC(=CC(=C1)N)C(=N)N.[Cl-].[Cl-]
C7H11Cl2N3
MFCD03790003
dihydrogen 3-aminobenzenecarboximidamide dichloride
3-aminobenzenecarboximidamide;hydron;dichloride

Specifications

Authentic
10% max. (K.F.)
500mg
97.5
98%
2HCl·xH2O
dihydrogen 3-aminobenzenecarboximidamide dichloride
[H+].[H+].C1=CC(=CC(=C1)N)C(=N)N.[Cl-].[Cl-]
208.086
208.09
≥97.5% (on dry substance)
Glass Bottle
Brown or Pink-White
37132-68-6
100.0
C7H11Cl2N3
MFCD03790003
FKFMMMVFFYPIEE-UHFFFAOYSA-N
3-aminobenzenecarboximidamide;hydron;dichloride
74764793
Powder
3-Aminobenzamidine dihydrochloride hydrate, 98%
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Harmful if swallowed.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 253-355-0

TSCA : TSCA

SDS