missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3′-Aminoacetophenone 98.0+%, TCI America™

$87.08 - $575.98

Chemical Identifiers

CAS 99-03-6
Molecular Formula C8H9NO
Molecular Weight (g/mol) 135.166
MDL Number MFCD00007796
InChI Key CKQHAYFOPRIUOM-UHFFFAOYSA-N
Synonym 3'-aminoacetophenone, 1-3-aminophenyl ethanone, m-aminoacetophenone, 3-aminoacetophenone, 3-acetylaniline, m-acetylaniline, ethanone, 1-3-aminophenyl, 1-3-aminophenyl ethan-1-one, m-aminoacetylbenzene, acetophenone, 3'-amino
PubChem CID 7417
IUPAC Name 1-(3-aminophenyl)ethanone
SMILES CC(=O)C1=CC(=CC=C1)N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
A024925G View Documents TCI America
A024925G
25g Each for $87.08
 
A0249500G View Documents TCI America
A0249500G
500g Each for $575.98
 
Specifications

Chemical Identifiers

99-03-6
135.166
CKQHAYFOPRIUOM-UHFFFAOYSA-N
7417
CC(=O)C1=CC(=CC=C1)N
C8H9NO
MFCD00007796
3'-aminoacetophenone, 1-3-aminophenyl ethanone, m-aminoacetophenone, 3-aminoacetophenone, 3-acetylaniline, m-acetylaniline, ethanone, 1-3-aminophenyl, 1-3-aminophenyl ethan-1-one, m-aminoacetylbenzene, acetophenone, 3'-amino
1-(3-aminophenyl)ethanone

Specifications

252°C
98°C
99-03-6
MFCD00007796
CKQHAYFOPRIUOM-UHFFFAOYSA-N
1-(3-aminophenyl)ethanone
7417
Crystal-Powder at 20°C
3′-Aminoacetophenone
Undesignated
25g
C8H9NO
3'-aminoacetophenone, 1-3-aminophenyl ethanone, m-aminoacetophenone, 3-aminoacetophenone, 3-acetylaniline, m-acetylaniline, ethanone, 1-3-aminophenyl, 1-3-aminophenyl ethan-1-one, m-aminoacetylbenzene, acetophenone, 3'-amino
CC(=O)C1=CC(=CC=C1)N
135.166
135.17
≥98.0% (GC,T)
Safety and Handling

Safety and Handling

EINECSNumber : (3)-1242

RTECSNumber : AM5800000

TSCA : Yes