Please login to your online account to display your discounted pricing

3'-Aminoacetophenone, 97%, ACROS Organics™

$15.48 - $15.48

Chemical Identifiers

Chemical Name or Material 3';-Aminoacetophenone
Name Note 97%
CAS 99-03-6
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C8H9NO
Linear Formula H2NC6H4COCH3
Formula Weight 135.17g/mol
MDL Number MFCD00007796
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC160770250

  SDS
fisher scientific acros
160770250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC160770050

  SDS
thermo fisher scientific
160770050
Glass bottle 5g Each for $15.48
Description & Specifications

Specifications

Packaging Glass bottle
Quantity 25g
Chemical Name or Material 3';-Aminoacetophenone
Name Note 97%
CAS 99-03-6
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C8H9NO
Linear Formula H2NC6H4COCH3
Formula Weight 135.17g/mol
MDL Number MFCD00007796
Beilstein 14, 45
Merck Index 14, 413
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
RTECS Number AM5800000
EINECS Number 202-722-3
TSCA TSCA
Safety and Handling

Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.