missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3'-Aminoacetanilide, 97%

Manufacturer:  Alfa Aesar™ H6062918

 View more versions of this product

Catalog No. AAH6062918



Description

Description

Applications
3′-Aminoacetanilide is mainly used as reaction intermediates dyes and disperse dyes.

Solubility
Soluble in water 1-5 g/100 mL at 24°C.

Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
Specifications

Specifications

3'-Aminoacetanilide
50g
C8H10N2O
n-3-aminophenyl acetamide, 3'-aminoacetanilide, 3-aminoacetanilide, m-aminoacetanilide, acetamide, n-3-aminophenyl, 3-acetamidoaniline, m-acetaminoaniline, m-acetylamino aniline, 3-aminoacetanilid, 3-amino-n-acetylaniline
CC(=O)NC1=CC=CC(=C1)N
150.181
150.18
86°C to 88°C
102-28-3
MFCD00025229
PEMGGJDINLGTON-UHFFFAOYSA-N
N-(3-aminophenyl)acetamide
7604
97%

Chemical Identifiers

102-28-3
150.181
PEMGGJDINLGTON-UHFFFAOYSA-N
7604
CC(=O)NC1=CC=CC(=C1)N
C8H10N2O
MFCD00025229
n-3-aminophenyl acetamide, 3'-aminoacetanilide, 3-aminoacetanilide, m-aminoacetanilide, acetamide, n-3-aminophenyl, 3-acetamidoaniline, m-acetaminoaniline, m-acetylamino aniline, 3-aminoacetanilid, 3-amino-n-acetylaniline
N-(3-aminophenyl)acetamide
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

RTECSNumber : AD8050000

TSCA : TSCA

Documents