missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3'-Aminoacetanilide, 97%

$37.47 - $149.89

Chemical Identifiers

CAS 102-28-3
Molecular Formula C8H10N2O
Molecular Weight (g/mol) 150.181
MDL Number MFCD00025229
InChI Key PEMGGJDINLGTON-UHFFFAOYSA-N
Synonym 3'-aminoacetanilide
PubChem CID 7604
IUPAC Name N-(3-aminophenyl)acetamide
SMILES CC(=O)NC1=CC=CC(=C1)N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6062918 View Documents Alfa Aesar™
H6062918
50g Each for $37.47
 
AAH6062930 View Documents Alfa Aesar™
H6062930
250g Each for $149.89
 
Description

Description

Applications
3′-Aminoacetanilide is mainly used as reaction intermediates dyes and disperse dyes.

Solubility
Soluble in water 1-5 g/100 mL at 24°C.

Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
Specifications

Chemical Identifiers

102-28-3
150.181
PEMGGJDINLGTON-UHFFFAOYSA-N
7604
CC(=O)NC1=CC=CC(=C1)N
C8H10N2O
MFCD00025229
3'-aminoacetanilide
N-(3-aminophenyl)acetamide

Specifications

86°C to 88°C
102-28-3
MFCD00025229
PEMGGJDINLGTON-UHFFFAOYSA-N
N-(3-aminophenyl)acetamide
7604
97%
50g
C8H10N2O
3'-aminoacetanilide
CC(=O)NC1=CC=CC(=C1)N
150.181
150.18
3'-Aminoacetanilide
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

RTECSNumber : AD8050000

TSCA : TSCA