Learn More
3-Amino-o-cresol, 95%, ACROS Organics™
Chemical Identifiers
CAS | 53222-92-7 |
---|---|
Molecular Formula | C7H9NO |
Molecular Weight (g/mol) | 123.15 |
MDL Number | MFCD00007787 |
InChI Key | FLROJJGKUKLCAE-UHFFFAOYSA-N |
Synonym | 2-amino-o-cresol, 2-methyl-3-amino-phenol, 2-methyl-3-aminophenol, 3-amino-2-methyl phenol, 3-amino-o-cresol, 3-hydroxy-2-methylaniline, aminokresol, o-cresol, 3-amino, phenol, 3-amino-2-methyl, pubchem15039 |
PubChem CID | 104448 |
IUPAC Name | 3-amino-2-methylphenol |
SMILES | CC1=C(C=CC=C1O)N |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC209042500
|
Thermo Fisher Scientific
209042500 |
250mg | Glass bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
53222-92-7 | |
123.15 | |
FLROJJGKUKLCAE-UHFFFAOYSA-N | |
104448 | |
CC1=C(C=CC=C1O)N |
C7H9NO | |
MFCD00007787 | |
2-amino-o-cresol, 2-methyl-3-amino-phenol, 2-methyl-3-aminophenol, 3-amino-2-methyl phenol, 3-amino-o-cresol, 3-hydroxy-2-methylaniline, aminokresol, o-cresol, 3-amino, phenol, 3-amino-2-methyl, pubchem15039 | |
3-amino-2-methylphenol |
Specifications
Glass bottle | |
250mg | |
94.0 | |
94% min. (GC) | |
H2NC6H3(CH3)OH | |
13, 579 | |
FLROJJGKUKLCAE-UHFFFAOYSA-N | |
3-amino-2-methylphenol | |
104448 | |
95% |
127°C to 133°C | |
53222-92-7 | |
100.0 | |
C7H9NO | |
MFCD00007787 | |
2-amino-o-cresol, 2-methyl-3-amino-phenol, 2-methyl-3-aminophenol, 3-amino-2-methyl phenol, 3-amino-o-cresol, 3-hydroxy-2-methylaniline, aminokresol, o-cresol, 3-amino, phenol, 3-amino-2-methyl, pubchem15039 | |
CC1=C(C=CC=C1O)N | |
123.15 | |
123.15 | |
3-Amino-o-cresol, 95% |
Safety and Handling
GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
EINECSNumber : 258-438-5
TSCA : TSCA