missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Amino-6-cyano-2-methylpyridine, 95%

$155.76 - $464.18

Chemical Identifiers

CAS 1079054-78-6
Molecular Formula C7H7N3
Molecular Weight (g/mol) 133.154
MDL Number MFCD08062828
InChI Key SLBIKHREAIGBFI-UHFFFAOYSA-N
Synonym 5-amino-6-methylpicolinonitrile, 6-cyano-2-methyl-3-aminopyridine, 5-amino-6-methyl-2-pyridinecarbonitrile, 5-amino-6-methyl-pyridine-2-carbonitrile
PubChem CID 55252984
IUPAC Name 5-amino-6-methylpyridine-2-carbonitrile
SMILES CC1=C(C=CC(=N1)C#N)N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH66325MD View Documents Alfa Aesar™
H66325MD
250mg Each for $155.76
 
AAH6632503 View Documents Alfa Aesar™
H6632503
1g Each for $464.18
 
Specifications

Chemical Identifiers

1079054-78-6
133.154
SLBIKHREAIGBFI-UHFFFAOYSA-N
55252984
CC1=C(C=CC(=N1)C#N)N
C7H7N3
MFCD08062828
5-amino-6-methylpicolinonitrile, 6-cyano-2-methyl-3-aminopyridine, 5-amino-6-methyl-2-pyridinecarbonitrile, 5-amino-6-methyl-pyridine-2-carbonitrile
5-amino-6-methylpyridine-2-carbonitrile

Specifications

250mg
C7H7N3
UN3439
5-amino-6-methylpicolinonitrile, 6-cyano-2-methyl-3-aminopyridine, 5-amino-6-methyl-2-pyridinecarbonitrile, 5-amino-6-methyl-pyridine-2-carbonitrile
CC1=C(C=CC(=N1)C#N)N
133.154
133.15
95%
1079054-78-6
MFCD08062828
15464648
SLBIKHREAIGBFI-UHFFFAOYSA-N
5-amino-6-methylpyridine-2-carbonitrile
55252984
Solid
3-Amino-6-cyano-2-methylpyridine
Safety and Handling

Safety and Handling

GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.