Please login to your online account to display your discounted pricing

3-Amino-6-chloropyridine, 98%, ACROS Organics™

$261.29 - $261.29

Chemical Identifiers

Chemical Name or Material 3-Amino-6-chloropyridine
CAS 5350-93-6
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C5H5ClN2
Formula Weight 128.56
MDL Number MFCD00006243
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC361330050

  SDS
thermo fisher scientific
AC361330050
Glass bottle 5g Each for $261.29

AC361330010

  SDS
thermo fisher scientific
AC361330010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥97.5%
Physical Form Powder, Crystals and/or Chunks
Formula Weight 128.56
Quantity 5g
Color Brown
Melting Point 80° to 84°C
CAS Max % 100.0
Assay Percent Range 98%
MDL Number MFCD00006243
CAS 5350-93-6
CAS Min % 97.5
Chemical Name or Material 3-Amino-6-chloropyridine
Packaging Glass bottle
Molecular Formula C5H5ClN2
Infrared Spectrum Authentic
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.