missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Amino-6-chloropyridazine, 98%

Manufacturer:  Alfa Aesar™ A1018503

 View more versions of this product

Catalog No. AAA1018503



Specifications

Specifications

3-Amino-6-chloropyridazine
1g
C4H4ClN3
110918
DTXVKPOKPFWSFF-UHFFFAOYSA-N
6-chloropyridazin-3-amine
21643
98%
~218°C (decomposition)
5469-69-2
MFCD00051506
3-amino-6-chloropyridazine, 3-pyridazinamine, 6-chloro, 6-chloro-3-pyridazinamine, 6-amino-3-chloropyridazine, 6-chloro-3-aminopyridazine, 6-chloropyridazin-3-ylamine, 6-chloro-pyridazin-3-ylamine, unii-ijs443slzg, 3-chloro-6-pyridazinamine, 6-chloro-3-pyridazinylamine
C1=CC(=NN=C1N)Cl
129.547
129.55

Chemical Identifiers

5469-69-2
129.547
DTXVKPOKPFWSFF-UHFFFAOYSA-N
21643
C1=CC(=NN=C1N)Cl
C4H4ClN3
MFCD00051506
3-amino-6-chloropyridazine, 3-pyridazinamine, 6-chloro, 6-chloro-3-pyridazinamine, 6-amino-3-chloropyridazine, 6-chloro-3-aminopyridazine, 6-chloropyridazin-3-ylamine, 6-chloro-pyridazin-3-ylamine, unii-ijs443slzg, 3-chloro-6-pyridazinamine, 6-chloro-3-pyridazinylamine
6-chloropyridazin-3-amine
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA

SDS Documents