missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Amino-6-chloro-5-methoxypyridine, 97%

$359.31 - $1,077.95

Chemical Identifiers

CAS 75711-01-2
Molecular Formula C6H7ClN2O
Molecular Weight (g/mol) 158.585
MDL Number MFCD09952112
InChI Key FJKVNEBFHWMVGF-UHFFFAOYSA-N
Synonym 3-amino-6-chloro-5-methoxy pyridine, 3-amino-6-chloro-5-methoxypyridine, 5-amino-2-chloro-3-methoxypyridine, 3-pyridinamine, 6-chloro-5-methoxy, pubchem22114, 6-chloro-5-methoxypyridine-3-amine, 6-chloro-5-methyloxy-3-pyridinamine, 3-amino-6-chloro-5-methoxypyridine
PubChem CID 23456143
IUPAC Name 6-chloro-5-methoxypyridin-3-amine
SMILES COC1=C(N=CC(=C1)N)Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH64033MD View Documents Alfa Aesar™
H64033MD
250mg Each for $359.31
 
AAH6403303 View Documents Alfa Aesar™
H6403303
1g Each for $1,077.95
 
Specifications

Chemical Identifiers

75711-01-2
158.585
FJKVNEBFHWMVGF-UHFFFAOYSA-N
23456143
COC1=C(N=CC(=C1)N)Cl
C6H7ClN2O
MFCD09952112
3-amino-6-chloro-5-methoxy pyridine, 3-amino-6-chloro-5-methoxypyridine, 5-amino-2-chloro-3-methoxypyridine, 3-pyridinamine, 6-chloro-5-methoxy, pubchem22114, 6-chloro-5-methoxypyridine-3-amine, 6-chloro-5-methyloxy-3-pyridinamine, 3-amino-6-chloro-5-methoxypyridine
6-chloro-5-methoxypyridin-3-amine

Specifications

250mg
C6H7ClN2O
15026636
FJKVNEBFHWMVGF-UHFFFAOYSA-N
6-chloro-5-methoxypyridin-3-amine
23456143
Solid
3-Amino-6-chloro-5-methoxypyridine
75711-01-2
MFCD09952112
3-amino-6-chloro-5-methoxy pyridine, 3-amino-6-chloro-5-methoxypyridine, 5-amino-2-chloro-3-methoxypyridine, 3-pyridinamine, 6-chloro-5-methoxy, pubchem22114, 6-chloro-5-methoxypyridine-3-amine, 6-chloro-5-methyloxy-3-pyridinamine, 3-amino-6-chloro-5-methoxypyridine
COC1=C(N=CC(=C1)N)Cl
158.585
158.59
97%