Please login to your online account to display your discounted pricing

3-amino-6-chloro-4-picoline, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC360990050

 View more versions of this product

Catalog No. AC360990050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 142.59
Physical Form Powder
Assay ≥97.5% (GC)
Quantity 5g
Color Bordeaux to brown
Melting Point 69° to 73°C
Assay Percent Range 98%
Chemical Name or Material 3-Amino-6-chloro-4-picoline
Infrared Spectrum Authentic
Packaging Glass bottle
CAS 66909-38-4
Molecular Formula C6H7ClN2

Specifications

Chemical Name or Material 3-Amino-6-chloro-4-picoline
Assay ≥97.5% (GC)
Assay Percent Range 98%
Molecular Formula C6H7ClN2
Formula Weight 142.59