Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Amino-6-chloro-2-methylpyridine, 95%

Chemical Identifiers

Chemical Name or Material 3-Amino-6-chloro-2-methylpyridine
CAS 164666-68-6
Formula Weight 142.59
MDL Number MFCD03095220
Synonym 3-Amino-6-chloro-2-picoline
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6146506

  SDS
alfa aesar™
H6146506
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6146503

  SDS
alfa aesar™
H6146503
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 142.59
Chemical Name or Material 3-Amino-6-chloro-2-methylpyridine
Color Pale yellow
Quantity 5g
Physical Form Solid
Assay 95%
MDL Number MFCD03095220
CAS 164666-68-6
Synonym 3-Amino-6-chloro-2-picoline
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H318-H302-H335-H315
Causes serious eye damage.
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Storage Note 1 -30° to -10°C
Safety and Handling

Danger

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H318-H302-H335-H315
Causes serious eye damage.
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.

-30° to -10°C