Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Amino-6-bromopyrazine-2-carboxylic acid, 98%

Chemical Identifiers

Chemical Name or Material 3-Amino-6-bromopyrazine-2-carboxylic acid
CAS 486424-37-7
Formula Weight 218.01
MDL Number MFCD08275681
Beilstein 147851
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6690803

  SDS
alfa aesar™
H6690803
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6690806

  SDS
alfa aesar™
H6690806
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Melting Point 178° to 183°C
Formula Weight 218.01
Chemical Name or Material 3-Amino-6-bromopyrazine-2-carboxylic acid
Physical Form Solid
MDL Number MFCD08275681
CAS 486424-37-7
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Assay 98%
Color Pale yellow
Quantity 1g
Beilstein 147851
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Sensitivity Light Sensitive
Storage Note 1 -30° to -10°C

Applications
3-Amino-6-bromopyrazine-2-carboxylic acid used to prepare 3-bromo-5-phenylpyrazine-2-amine. It serves as an intermediate in organic synthesis and pharmaceuticals.

Solubility
Soluble in dimethyl sulfoxide and methanol.

Notes
Incompatible with strong oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

-30° to -10°C