missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Amino-6-bromo-2-methylpyridine 97.0+%, TCI America™

$69.68 - $210.70

Chemical Identifiers

CAS 126325-47-1
Molecular Formula C6H7BrN2
Molecular Weight (g/mol) 187.04
MDL Number MFCD03095086
InChI Key UBTQJSZELCWGCN-UHFFFAOYSA-N
Synonym 5-amino-2-bromo-6-picoline, 3-amino-6-bromo-2-methylpyridine, 2-bromo-5-amino-6-methylpyridine, 3-amino-6-bromo-2-picoline, 5-amino-2-bromo-6-methylpyridine, 2-bromo-5-amino-6-picoline, 6-bromo-2-methyl-3-pyridylamine, 6-bromo-2-methyl-pyridin-3-ylamine, 2-methyl-3-amino-6-bromopyridine, pubchem1117
PubChem CID 14418045
IUPAC Name 6-bromo-2-methylpyridin-3-amine
SMILES CC1=C(N)C=CC(Br)=N1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
A29281G
View Documents Promotion Details
TCI America
A29281G
1 g
Each for $69.68
 
A29285G
View Documents Promotion Details
TCI America
A29285G
5 g
Each for $210.70
 
Description

Description

Specifications

Chemical Identifiers

126325-47-1
187.04
UBTQJSZELCWGCN-UHFFFAOYSA-N
14418045
CC1=C(N)C=CC(Br)=N1
C6H7BrN2
MFCD03095086
5-amino-2-bromo-6-picoline, 3-amino-6-bromo-2-methylpyridine, 2-bromo-5-amino-6-methylpyridine, 3-amino-6-bromo-2-picoline, 5-amino-2-bromo-6-methylpyridine, 2-bromo-5-amino-6-picoline, 6-bromo-2-methyl-3-pyridylamine, 6-bromo-2-methyl-pyridin-3-ylamine, 2-methyl-3-amino-6-bromopyridine, pubchem1117
6-bromo-2-methylpyridin-3-amine

Specifications

126325-47-1
C6H7BrN2
1 g
UBTQJSZELCWGCN-UHFFFAOYSA-N
6-bromo-2-methylpyridin-3-amine
14418045
≥97.0% (GC,T)
3-Amino-6-bromo-2-methylpyridine
110°C
MFCD03095086
5-amino-2-bromo-6-picoline, 3-amino-6-bromo-2-methylpyridine, 2-bromo-5-amino-6-methylpyridine, 3-amino-6-bromo-2-picoline, 5-amino-2-bromo-6-methylpyridine, 2-bromo-5-amino-6-picoline, 6-bromo-2-methyl-3-pyridylamine, 6-bromo-2-methyl-pyridin-3-ylamine, 2-methyl-3-amino-6-bromopyridine, pubchem1117
CC1=C(N)C=CC(Br)=N1
187.04
187.04
Crystalline Powder
Safety and Handling

Safety and Handling

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'