missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3-Amino-5-(trifluoromethyl)-1H-indazole, 95%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC435730050
Specifications
3-Amino-5-(trifluoromethyl)-1H-indazole | |
White to Yellow | |
5g | |
94.0 | |
94% min. (HPLC) | |
MFCD01719740 | |
ZXZTZRVCWTWKCH-UHFFFAOYSA-N | |
5-(trifluoromethyl)-1H-indazol-3-amine | |
16736 | |
Crystalline Powder |
Authentic | |
134.5°C to 135.5°C | |
2250-53-5 | |
100.0 | |
C8H6F3N3 | |
1h-indazol-3-amine, 5-trifluoromethyl, 1h-indazol-3-amine, 5-trifluoromethyl-9ci, 1h-indazole, 3-amino-5-trifluoromethyl, 3-amino-5-trifluoromethyl-1h-indazole, 3-amino-5-trifluoromethylindazole, 3-amino-5trifluoromethyl-indazole, 5-trifluoromethyl-1h-indazol-3-amine, 5-trifluoromethyl-1h-indazol-3-ylamine, 5-trifluoromethyl-1h-indazole-3-ylamine | |
C1=CC2=C(C=C1C(F)(F)F)C(=NN2)N | |
201.15 | |
201.15 | |
95% |
Chemical Identifiers
2250-53-5 | |
201.15 | |
ZXZTZRVCWTWKCH-UHFFFAOYSA-N | |
16736 | |
C1=CC2=C(C=C1C(F)(F)F)C(=NN2)N |
C8H6F3N3 | |
MFCD01719740 | |
1h-indazol-3-amine, 5-trifluoromethyl, 1h-indazol-3-amine, 5-trifluoromethyl-9ci, 1h-indazole, 3-amino-5-trifluoromethyl, 3-amino-5-trifluoromethyl-1h-indazole, 3-amino-5-trifluoromethylindazole, 3-amino-5trifluoromethyl-indazole, 5-trifluoromethyl-1h-indazol-3-amine, 5-trifluoromethyl-1h-indazol-3-ylamine, 5-trifluoromethyl-1h-indazole-3-ylamine | |
5-(trifluoromethyl)-1H-indazol-3-amine |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/e
GHS Signal Word: Warning