missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Amino-5-(trifluoromethyl)-1H-indazole, 95%, ACROS Organics™

Chemical Identifiers

CAS 2250-53-5
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 95%
Molecular Formula C8H6F3N3
Formula Weight 201.15
MDL Number MFCD01719740
Synonym 5-(Trifluoromethyl)-1H-indazol-3-ylamine
Chemical Name or Material 3-Amino-5-(trifluoromethyl)-1H-indazole
Assay ≥94.0%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC435730050 SDS Thermo Fisher Scientific
AC435730050
5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

2250-53-5
100.0
C8H6F3N3
MFCD01719740
3-Amino-5-(trifluoromethyl)-1H-indazole
94.0
95%
201.15
5-(Trifluoromethyl)-1H-indazol-3-ylamine
≥94.0%

Specifications

2250-53-5
100.0
Crystalline Powder
C8H6F3N3
201.15
MFCD01719740
5-(Trifluoromethyl)-1H-indazol-3-ylamine
≥94.0%
94.0
Authentic
95%
White to yellow
133° to 137°C
5g
3-Amino-5-(trifluoromethyl)-1H-indazole

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning