Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Amino-5-tert-butyl-1H-pyrazole, 98%

$108.34 - $1,652.92

Chemical Identifiers

Chemical Name or Material 3-Amino-5-tert-butyl-1H-pyrazole
CAS 82560-12-1
Molecular Formula C7H13N3
Formula Weight 139.2
MDL Number MFCD00067987
Synonym 5-tert-Butyl-1H-pyrazol-3-amine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1275003

  SDS
alfa aesar™
A1275003
1g Each for $108.34

AAA1275006

  SDS
alfa aesar™
A1275006
5g Each for $381.22

AAA1275014

  SDS
alfa aesar™
A1275014
25g Each for $1,652.92
Description & Specifications

Specifications

Formula Weight 139.2
Chemical Name or Material 3-Amino-5-tert-butyl-1H-pyrazole
Melting Point 75° to 79°C
Quantity 1g
Assay 98%
Molecular Formula C7H13N3
MDL Number MFCD00067987
CAS 82560-12-1
Synonym 5-tert-Butyl-1H-pyrazol-3-amine
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.