Please login to your online account to display your discounted pricing

3-Amino-5-tert-butyl-1H-pyrazole, 97%, Acros Organics

Chemical Identifiers

Chemical Name or Material 3-Amino-5-tert-butyl-1H-pyrazole
Assay Percent Range 97%
Molecular Formula C7H13N3
Formula Weight 139.2
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC448650050

  SDS
thermo fisher scientific
AC448650050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC448650500

  SDS
thermo fisher scientific
AC448650500
Glass bottle 50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 139.2
Quantity 5g
Chemical Name or Material 3-Amino-5-tert-butyl-1H-pyrazole
Assay Percent Range 97%
Molecular Formula C7H13N3
CAS 82560-12-1
Packaging Glass bottle