Please login to your online account to display your discounted pricing

3-Amino-5-tert-butyl-1H-pyrazole, 97%, Acros Organics


Quantity 50g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC448650500

 View more versions of this product

Catalog No. AC448650500

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 139.2
Quantity 50g
CAS 82560-12-1
Chemical Name or Material 3-Amino-5-tert-butyl-1H-pyrazole
Assay Percent Range 97%
Packaging Glass bottle
Molecular Formula C7H13N3

Specifications

Chemical Name or Material 3-Amino-5-tert-butyl-1H-pyrazole
Assay Percent Range 97%
Molecular Formula C7H13N3
Formula Weight 139.2