Please login to your online account to display your discounted pricing

3-Amino-5-nitrobenzoisothiazole, 95%, ACROS Organics™

$235.14 - $235.14

Chemical Identifiers

Chemical Name or Material 3-Amino-5-nitrobenzoisothiazole
CAS 84387-89-3
CAS Min % 94.0
CAS Max % 100.0
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C7H5N3O2S
Formula Weight 195.2
MDL Number MFCD00270828
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC345410250

  SDS
thermo fisher scientific
345410250
Glass bottle 25g Each for $235.14
Description & Specifications

Specifications

Assay ≥94%
Physical Form Powder
Formula Weight 195.2
Quantity 25g
Color Orange to red
CAS Min % 94.0
CAS Max % 100.0
Chemical Name or Material 3-Amino-5-nitrobenzoisothiazole
Assay Percent Range 95%
Molecular Formula C7H5N3O2S
MDL Number MFCD00270828
CAS 84387-89-3
Infrared Spectrum Authentic
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Packaging Glass bottle
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.