Please login to your online account to display your discounted pricing

3-Amino-5-methylpyrazole, 97%, ACROS Organics™


Quantity 100g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC368031000

 View more versions of this product

Catalog No. AC368031000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥96.0%
Formula Weight 97.12
Physical Form Solid
Quantity 100g
Color Light yellow to orange
Melting Point 39° to 44°C
CAS Min % 96.0
CAS Max % 100.0
Health Hazard 1 Warning
MDL Number MFCD00075180
Chemical Name or Material 3-Amino-5-methylpyrazole
Assay Percent Range 97%
Molecular Formula C4H7N3
CAS 31230-17-8
Name Note 97%
Packaging Glass bottle
Infrared Spectrum Authentic
Synonym 5-Methyl-3-pyrazolamine
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material 3-Amino-5-methylpyrazole
Name Note 97%
CAS 31230-17-8
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C4H7N3
Formula Weight 97.12
MDL Number MFCD00075180
Synonym 5-Methyl-3-pyrazolamine
Accessories

Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

SDS & Safety Documents