missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Amino-5-methyl-1H-pyrazole, 97%

Manufacturer:  Alfa Aesar™ A1164206

 View more versions of this product

Catalog No. AAA1164206


 

Description

Description

Applications
3-Amino-5-methyl-1H-pyrazole was employed as beta-sheet template to investigate its interaction with ferrocenoyl-dipeptides. It is also used in the synthesis of 4-aryl-3,7,7-trimethyl-2,4,6,7,8,9-hexahydropyrazolo[3,4-b]quinolin-5-ones. It can react with 4-Ethoxymethylene-2-phenyl-4H-oxazol-5-one to get N-(2-Methyl-7-oxo-4,7-dihydro-pyrazolo[1,5-a]pyrimidin-6-yl)-benzamide.

Solubility
Soluble in Dichloromethane and Methanol.

Notes
Air Sensitive. Store at -20°C. Store under dry inert gas away from oxidizing agents and air.
Specifications

Specifications

3-Amino-5-methyl-1H-pyrazole
97%
42°C to 47°C
212°C to 213°C (14mmHg)
5g
5-Methyl-1H-pyrazol-3-amine
CC1=CC(=NN1)N
97.121
97.12
31230-17-8
C4H7N3
MFCD00075180
1904
>110°C (230°F)
FYTLHYRDGXRYEY-UHFFFAOYSA-N
5-methyl-1H-pyrazol-3-amine
93146

Chemical Identifiers

31230-17-8
97.121
FYTLHYRDGXRYEY-UHFFFAOYSA-N
93146
C4H7N3
MFCD00075180
5-Methyl-1H-pyrazol-3-amine
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

SDS Documents