missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Amino-5-fluorobenzoic acid, 95%

$23.28 - $299.18

Chemical Identifiers

CAS 786616-54-4
Molecular Formula C7H6FNO2
Molecular Weight (g/mol) 155.128
MDL Number MFCD07779526
InChI Key RYLBYCHERDTVAY-UHFFFAOYSA-N
Synonym benzoic acid, 3-amino-5-fluoro-9ci, 3-amino-5-fluoro-benzoic acid, 5-amino-3-fluorobenzoic acid, 3-fluoro-5-aminobenzoic acid, benzoic acid,3-amino-5-fluoro, benzoic acid, 3-amino-5-fluoro
PubChem CID 17915148
IUPAC Name 3-amino-5-fluorobenzoic acid
SMILES C1=C(C=C(C=C1N)F)C(=O)O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH63501MD View Documents Alfa Aesar™
H63501MD
250mg Each for $23.28
 
AAH6350103 View Documents Alfa Aesar™
H6350103
1g Each for $68.93
 
AAH6350106 View Documents Alfa Aesar™
H6350106
5g Each for $299.18
 
Specifications

Chemical Identifiers

786616-54-4
155.128
RYLBYCHERDTVAY-UHFFFAOYSA-N
17915148
C1=C(C=C(C=C1N)F)C(=O)O
C7H6FNO2
MFCD07779526
benzoic acid, 3-amino-5-fluoro-9ci, 3-amino-5-fluoro-benzoic acid, 5-amino-3-fluorobenzoic acid, 3-fluoro-5-aminobenzoic acid, benzoic acid,3-amino-5-fluoro, benzoic acid, 3-amino-5-fluoro
3-amino-5-fluorobenzoic acid

Specifications

250mg
C7H6FNO2
benzoic acid, 3-amino-5-fluoro-9ci, 3-amino-5-fluoro-benzoic acid, 5-amino-3-fluorobenzoic acid, 3-fluoro-5-aminobenzoic acid, benzoic acid,3-amino-5-fluoro, benzoic acid, 3-amino-5-fluoro
C1=C(C=C(C=C1N)F)C(=O)O
155.128
155.13
95%
786616-54-4
MFCD07779526
RYLBYCHERDTVAY-UHFFFAOYSA-N
3-amino-5-fluorobenzoic acid
17915148
Solid
3-Amino-5-fluorobenzoic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

storageNote1 : -30° to -10°C